Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WBSCR22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202560.2 | NP_001189489.1 | 10 | 726 | ||
NM_017528.4 | NP_059998.2 | 10 | 726 | |||
NR_037776.2 | 10 | 922 | ||||
NR_045512.1 | 9 | 629 | ||||
XM_006715847.1 | XP_006715910.1 | 10 | 819 | |||
GenBank mRNA | AF218007.1 | - | 9 | 620 | ||
AF412034.1 | - | 10 | 726 | |||
AF420248.1 | - | 10 | 704 | |||
AJ224442.1 | - | 9 | 1367 | |||
AK091162.1 | - | 8 | 590 | |||
AK291116.1 | - | 10 | 697 | |||
AK296375.1 | - | 9 | 626 | |||
AK309574.1 | - | 8 | 527 | |||
AK315032.1 | - | 10 | 723 | |||
BC000169.2 | - | 10 | 682 | 2901188 | ||
BC001780.1 | - | 10 | 705 | 3544156 | ||
BC011696.2 | - | 10 | 687 | 3533955 | ||
BG108222.1 | - | 9 | 574 | |||
BM794274.1 | - | 5 | 316 | |||
BQ927478.1 | - | 667 | ||||
BQ930813.1 | - | 1 | 29 |