Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000167.5 | NP_000158.1 | 11 | 1031 | ||
NM_001128127.2 | NP_001121599.1 | 11 | 1031 | |||
NM_001205019.1 | NP_001191948.1 | 12 | 1049 | |||
NM_203391.3 | NP_976325.1 | 12 | 1049 | |||
XM_005274488.4 | XP_005274545.1 | 9 | 801 | |||
XM_006724483.2 | XP_006724546.1 | 12 | 1068 | |||
XM_006724484.2 | XP_006724547.1 | 12 | 1067 | |||
XM_006724485.2 | XP_006724548.1 | 11 | 894 | |||
XM_006724486.3 | XP_006724549.1 | 10 | 879 | |||
XM_011545491.2 | XP_011543793.1 | 13 | 1087 | |||
XM_011545492.1 | XP_011543794.1 | 13 | 1086 | |||
XM_011545493.2 | XP_011543795.1 | 11 | 769 | |||
XM_011545494.2 | XP_011543796.1 | 11 | 714 | |||
XM_017029409.1 | XP_016884898.1 | 13 | 1084 | |||
XM_017029410.1 | XP_016884899.1 | 10 | 863 | |||
XM_017029411.1 | XP_016884900.1 | 10 | 878 | |||
XM_017029412.1 | XP_016884901.1 | 9 | 799 | |||
GenBank mRNA | AK294304.1 | - | 12 | 855 | ||
AK294938.1 | - | 13 | 1084 | |||
AK308238.1 | - | 9 | 710 | |||
AK313215.1 | - | 11 | 967 | |||
BC037549.1 | - | 11 | 888 | 5213716 | ||
BC042421.1 | - | 12 | 1050 | 4820945 | ||
BC071595.1 | - | 12 | 1050 | 5276056 | ||
BX647604.1 | - | 12 | 870 | |||
L13943.1 | - | 11 | 918 | |||
X68285.1 | - | 10 | 642 | |||
X69886.1 | - | 7 | 467 |
Gene Symbol: GK3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026575.1 | 1 | 1230 | |||
GenBank mRNA | AK225740.1 | - | 1 | 881 | ||
AK292282.1 | - | 1 | 1230 | |||
BC066960.1 | - | 1 | 886 |