Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134422.1 | NP_001127894.1 | 1, 2 | 703 | ||
NM_017548.4 | NP_060018.1 | 1, 2 | 703 | |||
XM_005247588.4 | XP_005247645.1 | 1, 2 | 730 | |||
XM_005247589.4 | XP_005247646.1 | 1, 2 | 730 | |||
XM_005247590.4 | XP_005247647.1 | 1, 2 | 730 | |||
XM_005247591.4 | XP_005247648.1 | 1, 2 | 730 | |||
XM_005247592.4 | XP_005247649.1 | 1, 2 | 204 | |||
XM_005247594.4 | XP_005247651.1 | 1, 2 | 111 | |||
XM_011512961.2 | XP_011511263.1 | 1, 2 | 730 | |||
GenBank mRNA | AF103803.1 | - | 1, 2 | 571 | ||
BC007338.2 | - | 1, 2 | 317 | 3619262 | ||
DB455474.1 | - | 1, 2 | 255 |