Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYDIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198542.1 | NP_001185471.1 | 2 | 229 | ||
NM_001198543.1 | NP_001185472.1 | 2 | 199 | |||
NM_001270974.2 | NP_001257903.1 | 2 | 297 | |||
NM_017558.4 | NP_060028.2 | 2 | 297 | |||
XM_006721206.2 | XP_006721269.1 | 2 | 191 | |||
XM_011523146.2 | XP_011521448.1 | 2 | 224 | |||
XM_011523147.1 | XP_011521449.1 | 2 | 191 | |||
XM_011523148.1 | XP_011521450.1 | 2 | 264 | |||
XM_011523151.1 | XP_011521453.1 | 2 | 223 | |||
XM_017023346.1 | XP_016878835.1 | 2 | 469 | |||
GenBank mRNA | AK022933.1 | - | 2 | 231 | ||
AK299016.1 | - | 2 | 186 | |||
AK299348.1 | - | 2 | 229 | |||
AK308296.1 | - | 1 | 16 | |||
AL705531.1 | - | 2 | 241 | |||
BC028351.1 | - | 2 | 178 | |||
BP228881.1 | - | 2 | 216 | |||
DA758908.1 | - | 2 | 264 | |||
DB079205.1 | - | 2 | 264 |