Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017583.5 | NP_060053.2 | 1 | 602 | ||
XM_006718254.1 | XP_006718317.1 | 1 | 662 | |||
GenBank mRNA | AJ249128.1 | - | 1 | 419 | ||
AK074242.1 | - | 1 | 426 | |||
BC013166.1 | - | 1 | 549 | 3924490 | ||
BC024031.2 | - | 1 | 449 | 3532065 | ||
BI550863.1 | - | 1 | 607 |