Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145099.1 | NP_001138571.1 | 9 | 1303 | ||
NM_017585.3 | NP_060055.2 | 10 | 1489 | |||
XM_011518189.2 | XP_011516491.2 | 10 | 1466 | |||
XM_017014237.1 | XP_016869726.1 | 10 | 1457 | |||
XM_017014238.1 | XP_016869727.1 | 10 | 1461 | |||
GenBank mRNA | AB209108.1 | - | 4 | 4722 | ||
AJ011372.2 | - | 10 | 1431 | |||
AK074836.1 | - | 9 | 1253 | |||
AK074927.1 | - | 10 | 1472 | |||
AK096630.1 | - | 8 | 2366 | |||
AK222919.1 | - | 10 | 1434 | |||
AK225106.1 | - | 10 | 1362 | |||
BC013740.1 | - | 10 | 1423 | 4449594 | ||
Y17803.1 | - | 10 | 1440 |