Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135775.2 | NP_001129247.1 | 4 | 1385 | ||
NM_001242369.1 | NP_001229298.1 | 6 | 1636 | |||
NM_001242370.1 | NP_001229299.1 | 5 | 1510 | |||
NM_017586.3 | NP_060056.1 | 5 | 1511 | |||
XM_011518181.2 | XP_011516483.1 | 6 | 3193 | |||
XM_011518182.1 | XP_011516484.1 | 6 | 1689 | |||
XM_011518184.1 | XP_011516486.1 | 5 | 2252 | |||
XM_011518185.2 | XP_011516487.1 | 5 | 3068 | |||
XM_011518186.1 | XP_011516488.1 | 5 | 1564 | |||
GenBank mRNA | AJ011373.1 | - | 5 | 1467 | ||
AK074852.1 | - | 5 | 1488 | |||
AK075416.1 | - | 4 | 1383 | |||
AK075548.1 | - | 5 | 1478 | |||
AK311325.1 | - | 5 | 1567 | |||
AL137319.1 | - | 1 | 56 | |||
AY358452.1 | - | 5 | 1459 | |||
BC030558.1 | - | 5 | 1477 | 5223979 |