Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017612.4 | NP_060082.2 | 10 | 1120 | ||
XM_006719491.2 | XP_006719554.1 | 7 | 805 | |||
XM_011538555.2 | XP_011536857.1 | 9 | 1662 | |||
XM_017019606.1 | XP_016875095.1 | 10 | 1040 | |||
GenBank mRNA | AK027702.1 | - | 10 | 989 | ||
AK074638.1 | - | 10 | 1018 | |||
AK291174.1 | - | 6 | 455 | |||
AL157433.1 | - | 8 | 2529 | |||
BC017704.2 | - | 3 | 244 | |||
BC065918.1 | - | 10 | 959 | 5752053 |