Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017625.2 | NP_060095.2 | 3 | 250 | ||
GenBank mRNA | AB036706.1 | - | 3 | 162 | ||
AF271386.1 | - | 3 | 173 | |||
AK000029.1 | - | 3 | 199 | |||
AK222533.1 | - | 3 | 237 | |||
AY065972.1 | - | 3 | 240 | |||
AY157361.1 | - | 3 | 252 | |||
AY157362.1 | - | 3 | 244 | |||
AY358359.1 | - | 3 | 236 | |||
AY549722.1 | - | 3 | 339 | |||
AY619691.1 | - | 3 | 160 | |||
AY619692.1 | - | 3 | 217 | |||
BC020664.1 | - | 3 | 250 | 4693329 | ||
CR457224.1 | - | 2 | 134 |