Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1RAPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017416.1 | NP_059112.1 | 5 | 1422 | ||
XM_011530905.2 | XP_011529207.1 | 3 | 430 | |||
XM_017029403.1 | XP_016884892.1 | 5 | 840 | |||
GenBank mRNA | AF181285.1 | - | 3 | 584 | ||
AF212016.1 | - | 4 | 666 | |||
AF284436.1 | - | 5 | 817 | |||
AJ272208.1 | - | 5 | 1422 | |||
AJ290436.1 | - | 4 | 685 | |||
AK290196.1 | - | 5 | 1538 | |||
BC104784.1 | - | 5 | 714 | 8143787 | ||
BC104786.1 | - | 5 | 714 | 8143789 |