Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPA17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017425.3 | NP_059121.1 | 5 | 890 | ||
XM_011542870.2 | XP_011541172.1 | 5 | 1080 | |||
GenBank mRNA | AF334735.1 | - | 5 | 889 |