Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278603.1 | NP_001265532.1 | 11 | 2005 | ||
NM_017426.3 | NP_059122.2 | 12 | 2149 | |||
NR_103780.1 | 11 | 1922 | ||||
NR_103781.1 | 10 | 1814 | ||||
NR_103782.1 | 12 | 1919 | ||||
XM_006714237.1 | XP_006714300.1 | 13 | 2126 | |||
XM_011532033.2 | XP_011530335.1 | 11 | 1984 | |||
XM_011532034.2 | XP_011530336.1 | 11 | 2392 | |||
XM_017008294.1 | XP_016863783.1 | 12 | 2536 | |||
XM_017008295.1 | XP_016863784.1 | 11 | 2309 | |||
GenBank mRNA | AF157322.1 | - | 12 | 2027 | ||
AK001517.1 | - | 12 | 2030 | |||
AL137650.1 | - | 4 | 1448 | |||
BC012559.1 | - | 12 | 2046 | |||
BX538002.1 | - | 10 | 1714 |