Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005112.4 | NP_005103.2 | 12 | 2500 | ||
NM_017491.3 | NP_059830.1 | 15 | 2920 | |||
XM_017008880.1 | XP_016864369.1 | 15 | 3113 | |||
GenBank mRNA | AB209746.1 | - | 15 | 2693 | ||
AF020056.1 | - | 15 | 2748 | |||
AF020260.1 | - | 15 | 2320 | |||
AK127665.1 | - | 15 | 2660 | |||
AL050108.1 | - | 13 | 2468 | |||
AL110208.1 | - | 6 | 1522 | |||
BC000201.1 | - | 15 | 2719 | 3351638 | ||
BC002489.2 | - | 15 | 2711 | 3051319 | ||
EU794675.1 | - | 15 | 2711 | |||
L32559.1 | - | 1 | 104 |