Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198321.3 | NP_938080.1 | 12 | 2363 | ||
GenBank mRNA | AK023782.1 | - | 9 | 1697 | ||
AK023815.1 | - | 3 | 813 | |||
AK127135.1 | - | 12 | 2332 | |||
AL096739.1 | - | 5 | 1507 | |||
BC007224.1 | - | 5 | 1163 | |||
BC072450.1 | - | 8 | 1663 | |||
BC098444.1 | - | 7 | 1352 |