Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POGK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017542.4 | NP_060012.3 | 3 | 441 | ||
XM_005245370.3 | XP_005245427.1 | 3 | 562 | |||
XM_011509814.2 | XP_011508116.1 | 3 | 718 | |||
XM_011509815.1 | XP_011508117.1 | 3 | 373 | |||
XM_017001914.1 | XP_016857403.1 | 3 | 435 | |||
GenBank mRNA | AB040946.1 | - | 3 | 358 | ||
AK056711.1 | - | 3 | 339 | |||
BC131624.1 | - | 3 | 326 | 40117695 | ||
DB049597.1 | - | 3 | 441 |