Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017583.5 | NP_060053.2 | 5 | 2175 | ||
XM_006718254.1 | XP_006718317.1 | 4 | 1995 | |||
GenBank mRNA | AK021823.1 | - | 5 | 1281 | ||
AK074242.1 | - | 5 | 1999 | |||
BC024031.2 | - | 5 | 2022 | 3532065 |