Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135775.2 | NP_001129247.1 | 4 | 1720 | ||
NM_001242369.1 | NP_001229298.1 | 6 | 1971 | |||
NM_001242370.1 | NP_001229299.1 | 5 | 1845 | |||
NM_017586.3 | NP_060056.1 | 5 | 1846 | |||
XM_011518181.2 | XP_011516483.1 | 6 | 3528 | |||
XM_011518182.1 | XP_011516484.1 | 6 | 2024 | |||
XM_011518184.1 | XP_011516486.1 | 5 | 2587 | |||
XM_011518185.2 | XP_011516487.1 | 5 | 3403 | |||
XM_011518186.1 | XP_011516488.1 | 5 | 1899 | |||
GenBank mRNA | AJ011373.1 | - | 5 | 1802 | ||
AK074852.1 | - | 5 | 1823 | |||
AK075416.1 | - | 4 | 1718 | |||
AK075548.1 | - | 5 | 1813 | |||
AL137319.1 | - | 1 | 391 | |||
AY358452.1 | - | 5 | 1794 | |||
BC030558.1 | - | 5 | 1812 | 5223979 |