Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKIRAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001349.2 | NP_001001349.1 | 4 | 1085 | ||
NM_001144927.1 | NP_001138399.1 | 4 | 1189 | |||
NM_001144928.1 | NP_001138400.1 | 4 | 1002 | |||
NM_001144929.1 | NP_001138401.1 | 5 | 1073 | |||
NM_017595.5 | NP_060065.2 | 4 | 1170 | |||
XM_005257251.1 | XP_005257308.1 | 4 | 1197 | |||
GenBank mRNA | AK027265.1 | - | 5 | 1042 | ||
AK054571.1 | - | 4 | 1197 | |||
AK297975.1 | - | 4 | 970 | |||
AL137682.1 | - | 2 | 2910 | |||
BC007316.2 | - | 4 | 1122 | |||
BC007450.2 | - | 4 | 1078 | 3028708 | ||
BC063498.1 | - | 4 | 1641 | 5248950 |