Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198950.1 | NP_001185879.1 | 27, 28 | 3954 | ||
NM_015011.1 | NP_055826.1 | 27, 28 | 3392 | |||
XM_011521062.1 | XP_011519364.1 | 27, 28 | 3500 | |||
GenBank mRNA | AB020672.2 | - | 27, 28 | 3392 | ||
AB290159.1 | - | 27, 28 | 3955 | |||
AK302933.1 | - | 15, 16 | 2095 | |||
BC146791.1 | - | 27, 28 | 3392 | 9007256 |