Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001264573.1 | NP_001251503.1 | 13 | 2216 | ||
NM_001265577.1 | NP_001252506.1 | 13 | 2180 | |||
XM_011524385.2 | XP_011522687.1 | 13 | 2517 | |||
XM_011524386.2 | XP_011522688.1 | 13 | 2251 | |||
XM_011524387.2 | XP_011522689.1 | 13 | 2491 | |||
XM_011524388.2 | XP_011522690.1 | 13 | 2481 | |||
XM_011524389.2 | XP_011522691.1 | 13 | 2517 | |||
XM_011524390.2 | XP_011522692.1 | 12 | 2413 | |||
GenBank mRNA | BC044933.1 | - | 4 | 849 | ||
BC067365.1 | - | 1 | 379 | |||
BC136590.1 | - | 12 | 2106 | 9020580 | ||
BC144271.1 | - | 12 | 2142 | 9052791 | ||
GU808359.1 | - | 12 | 2145 |