Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080467.2 | NP_001073936.1 | 26 | 3767 | ||
GenBank mRNA | AB032945.2 | - | 26 | 3497 | ||
AB290160.1 | - | 26 | 3587 | |||
AK025336.1 | - | 6 | 910 | |||
AL133643.1 | - | 5 | 544 |