Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017612.4 | NP_060082.2 | 14 | 2301 | ||
XM_006719491.2 | XP_006719554.1 | 11 | 1986 | |||
XM_011538555.2 | XP_011536857.1 | 13 | 2843 | |||
XM_017019606.1 | XP_016875095.1 | 14 | 2221 | |||
GenBank mRNA | AK026831.1 | - | 3 | 1170 | ||
AK027702.1 | - | 14 | 2170 | |||
AK074638.1 | - | 14 | 2199 | |||
AK291174.1 | - | 10 | 1636 | |||
AL157433.1 | - | 12 | 3710 | |||
BC065918.1 | - | 14 | 2140 | 5752053 |