Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDE9A SIRNA Assays ›
Gene Symbol:
PDE9A
Gene Name:
phosphodiesterase 9A
Gene Aliases:
HSPDE9A2
Chromosome Location:
Chr.21: 42653752 - 42775509 on Build GRCh38
UniGene ID:
Hs.473927
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
251301
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDE9A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001567.1
NP_001001567.1
15
1285
NM_001001568.1
NP_001001568.1
13
1136
NM_001001569.1
NP_001001569.1
12
1081
NM_001001570.1
NP_001001570.1
15
1306
NM_001001571.1
NP_001001571.1
14
1214
NM_001001572.1
NP_001001572.1
12
1092
NM_001001573.1
NP_001001573.1
13
1272
NM_001001574.1
NP_001001574.1
14
1207
NM_001001575.1
NP_001001575.1
11
1037
NM_001001576.1
NP_001001576.1
11
996
NM_001001577.1
NP_001001577.1
12
1115
NM_001001578.1
NP_001001578.1
13
1159
NM_001001579.1
NP_001001579.1
13
1152
NM_001001580.1
NP_001001580.1
10
952
NM_001001581.1
NP_001001581.1
14
1316
NM_001001582.1
NP_001001582.1
15
1394
NM_001001583.1
NP_001001583.1
15
1387
NM_001001584.2
NP_001001584.1
14
1215
NM_001001585.1
NP_001001585.1
14
1339
NM_001315533.1
NP_001302462.1
16
1361
NM_002606.2
NP_002597.1
16
1465
XM_011529598.2
XP_011527900.1
16
1387
XM_011529600.2
XP_011527902.1
17
1616
XM_017028366.1
XP_016883855.1
14
1203
GenBank mRNA
AF048837.1
-
16
1462
AF067223.1
-
16
1414
AF067224.1
-
15
1233
AF067225.1
-
13
1075
AF067226.1
-
12
1020
AK001165.1
-
13
1067
AK001307.1
-
15
1256
AK127770.1
-
6
2816
AK294112.1
-
13
1102
AK314679.1
-
16
1438
AY196299.1
-
15
1279
AY196300.1
-
14
1187
AY196301.1
-
12
1065
AY196302.1
-
13
1245
AY196303.1
-
14
1180
AY196304.1
-
11
1010
AY196305.1
-
11
969
AY196306.1
-
12
1088
AY196307.1
-
13
1132
AY196308.1
-
13
1125
AY196309.1
-
10
925
AY196310.1
-
14
1289
AY196311.1
-
15
1367
AY196312.1
-
15
1360
AY196313.1
-
14
1187
AY196314.1
-
14
1312
AY242121.1
-
14
1151
AY701187.1
-
16
1358
BC009047.1
-
15
1259
3874635
BT007016.1
-
15
1222
KU178246.1
-
15
1222
KU178247.1
-
16
1402
KU178248.1
-
12
1018
Back To Top
More Information
Panther Classification:
Molecular Function -
phosphodiesterase
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
positive regulation of cardiac muscle hypertrophy
cGMP-mediated signaling
cGMP metabolic process
cGMP catabolic process
3',5'-cyclic-nucleotide phosphodiesterase activity
protein binding
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline