Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004105.2 | NP_001004105.1 | 15 | 1804 | ||
NM_001004106.2 | NP_001004106.1 | 15 | 1804 | |||
NM_002082.3 | NP_002073.2 | 15 | 1804 | |||
XM_006714858.3 | XP_006714921.1 | 13 | 2445 | |||
XM_006714859.2 | XP_006714922.1 | 15 | 1716 | |||
XM_011534537.2 | XP_011532839.1 | 14 | 1556 | |||
XM_011534538.1 | XP_011532840.1 | 8 | 998 | |||
GenBank mRNA | AF040751.1 | - | 15 | 1647 | ||
AF040752.1 | - | 15 | 1647 | |||
AK056697.1 | - | 13 | 1782 | |||
BC009277.2 | - | 15 | 1744 | 4053197 | ||
BC017272.1 | - | 15 | 1693 | 4640283 | ||
L16862.1 | - | 15 | 1706 |