Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hps5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005247.2 | NP_001005247.2 | 23 | 3988 | ||
NM_001005248.2 | NP_001005248.2 | 23 | 3964 | |||
NM_001167864.1 | NP_001161336.1 | 23 | 3889 | |||
GenBank mRNA | AF534396.1 | - | 23 | 3785 | ||
AF534397.1 | - | 23 | 3761 | |||
AF534398.1 | - | 21 | 3350 | |||
AF534399.1 | - | 24 | 3916 | |||
AK129265.1 | - | 23 | 3911 | |||
AK140780.1 | - | 12 | 2170 | |||
AK141088.1 | - | 22 | 4614 | |||
AK157182.1 | - | 23 | 3754 | |||
AK161039.1 | - | 14 | 2359 | |||
BC046405.1 | - | 18 | 2967 | |||
BC046443.1 | - | 23 | 3602 | |||
BC082542.1 | - | 23 | 3889 | 6405811 |