Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013110.1 | NP_001013128.1 | 2 | 224 | ||
GenBank mRNA | AF476964.1 | - | 2 | 255 | ||
AY327504.1 | - | 2 | 193 | |||
BC087021.1 | - | 2 | 224 | 7132382 | ||
D38380.1 | - | 2 | 223 | |||
X77158.1 | - | 2 | 210 |
Gene Symbol: Srprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY325214.1 | - | 233 | |||
AY325230.1 | - | 233 | ||||
AY325241.1 | - | 233 |