Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd3b6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017265.4 | NP_058961.4 | 2, 3 | 373 | ||
GenBank mRNA | BC089937.1 | - | 2, 3 | 373 | 7309637 | |
L17138.1 | - | 1, 2 | 211 |
Gene Symbol: LOC100911154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749361.2 | XP_003749409.1 | 2, 3 | 392 |