Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cgnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK164015.1 | - | 1 | 694 | ||
AK173252.1 | - | 17 | 4825 | |||
BC026522.1 | - | 1 | 1140 | |||
BC031499.1 | - | 3 | 2435 | |||
BC038477.1 | - | 1 | 990 | |||
BC039211.1 | - | 11 | 3543 | |||
BC108380.1 | - | 1 | 1147 | |||
BC139006.1 | - | 18 | 5961 | 8862028 | ||
BC158061.1 | - | 18 | 5971 | 9007361 |