Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nnat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270867.1 | NP_001257796.1 | 3 | 482 | ||
NM_053601.2 | NP_446053.1 | 3 | 536 | |||
NM_181687.2 | NP_859015.1 | 2 | 455 | |||
XM_008762390.1 | XP_008760612.1 | 3 | 526 | |||
GenBank mRNA | BC127473.1 | - | 2 | 389 | 8364201 | |
CB711943.1 | - | 3 | 472 | |||
DY315493.1 | - | 3 | 487 | |||
FM037467.1 | - | 3 | 482 | |||
FQ213121.1 | - | 3 | 494 | |||
FQ213576.1 | - | 2 | 413 | |||
U08290.1 | - | 3 | 455 | |||
U09785.1 | - | 2 | 374 |