Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159651.1 | NP_001153123.1 | 3 | 1160 | ||
NM_138369.2 | NP_612378.1 | 4 | 1357 | |||
XM_017010017.1 | XP_016865506.1 | 2 | 1449 | |||
GenBank mRNA | AK301982.1 | - | 4 | 1345 | ||
BC003114.1 | - | 3 | 1085 | 3537973 | ||
BC007436.2 | - | 4 | 1347 | 2961368 | ||
DA506589.1 | - | 1 | 15 |