Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Miox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145771.2 | NP_665714.2 | 4 | 269 | ||
XM_008765674.1 | XP_008763896.1 | 4 | 453 | |||
GenBank mRNA | AF197128.1 | - | 4 | 263 | ||
AF230096.1 | - | 3 | 271 | |||
AY738259.1 | - | 3 | 263 | |||
BC078840.1 | - | 4 | 269 | 7110224 | ||
FQ219187.1 | - | 3 | 258 |