Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrgprx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145787.2 | NP_665730.2 | 1, 2 | 73 | ||
GenBank mRNA | AJ311952.1 | - | 19 | |||
BC089774.1 | - | 1, 2 | 73 | 7321342 |
Set Membership: |
G-protein coupled receptor |