Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM46A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017633.2 | NP_060103.2 | 2 | 740 | ||
GenBank mRNA | AF350451.1 | - | 2 | 701 | ||
AJ420592.1 | - | 2 | 575 | |||
AK000044.1 | - | 2 | 723 | |||
AK056057.1 | - | 2 | 807 | |||
AK292109.1 | - | 2 | 741 | |||
AY740520.1 | - | 1 | 437 | |||
BC000683.2 | - | 2 | 747 | 3349780 | ||
BC007351.2 | - | 2 | 739 | 3615536 | ||
BX648876.1 | - | 2 | 391 | |||
FJ211061.1 | - | 1 | 407 |