Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178912.3 | NP_849243.2 | 3 | 812 | ||
GenBank mRNA | AK039944.1 | - | 3 | 816 | ||
AK050306.1 | - | 3 | 813 | |||
AK053715.1 | - | 3 | 812 | |||
AK084697.1 | - | 3 | 816 | |||
AK086395.1 | - | 3 | 815 | |||
AK173211.1 | - | 3 | 791 | |||
BC150785.1 | - | 3 | 806 | 9087696 |