Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270890.1 | NP_001257819.1 | 9 | 1247 | ||
NM_017645.4 | NP_060115.3 | 9 | 1247 | |||
XM_017014830.1 | XP_016870319.1 | 5 | 398 | |||
GenBank mRNA | AB046794.1 | - | 2 | 103 | ||
AF533709.1 | - | 9 | 1429 | |||
AF537091.1 | - | 9 | 1209 | |||
AK022964.1 | - | 6 | 563 | |||
AK023068.1 | - | 2 | 166 | |||
AK056458.1 | - | 9 | 1209 | |||
AK301852.1 | - | 2 | 107 | |||
AL832495.1 | - | 9 | 1207 | |||
BC026178.1 | - | 9 | 1210 | |||
BC065935.1 | - | 7 | 747 | |||
BC071625.1 | - | 2 | 206 | |||
BC111041.1 | - | 5 | 523 | 5529130 | ||
BC113981.1 | - | 9 | 1044 | 40082296 | ||
BC114492.1 | - | 9 | 1044 | 40082291 |