Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr495 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000310.1 | NP_001000310.1 | 1 | 792 |
Gene Symbol: LOC100909940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749508.2 | XP_003749556.1 | 1 | 792 |