Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTSJ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017647.3 | NP_060117.3 | 4 | 834 | ||
GenBank mRNA | AF327355.1 | - | 3 | 309 | ||
AK095861.1 | - | 4 | 834 | |||
AK296715.1 | - | 4 | 233 | |||
AK315010.1 | - | 4 | 259 | |||
BC036710.2 | - | 3 | 218 | 4641078 |