Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001519.1 | NP_001001519.1 | 5 | 496 | ||
GenBank mRNA | AF303086.1 | - | 5 | 496 |