Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPATCH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002909.3 | NP_001002909.1 | 8 | 1240 | ||
NM_001304939.1 | NP_001291868.1 | 7 | 1165 | |||
NM_001304940.1 | NP_001291869.1 | 10 | 1381 | |||
NM_001304941.1 | NP_001291870.1 | 9 | 1258 | |||
NM_001304942.1 | NP_001291871.1 | 7 | 1167 | |||
NM_001304943.1 | NP_001291872.1 | 5 | 1121 | |||
XM_011524557.1 | XP_011522859.1 | 10 | 1314 | |||
XM_011524558.1 | XP_011522860.1 | 11 | 1133 | |||
XM_011524559.2 | XP_011522861.1 | 12 | 1211 | |||
XM_011524561.2 | XP_011522863.1 | 8 | 972 | |||
XM_017024372.1 | XP_016879861.1 | 13 | 1256 | |||
XM_017024373.1 | XP_016879862.1 | 10 | 2091 | |||
XM_017024374.1 | XP_016879863.1 | 12 | 1174 | |||
XM_017024375.1 | XP_016879864.1 | 9 | 1269 | |||
XM_017024376.1 | XP_016879865.1 | 10 | 1129 | |||
XM_017024377.1 | XP_016879866.1 | 9 | 1106 | |||
XM_017024378.1 | XP_016879867.1 | 10 | 1287 | |||
XM_017024379.1 | XP_016879868.1 | 11 | 1169 | |||
XM_017024380.1 | XP_016879869.1 | 10 | 1107 | |||
GenBank mRNA | AB011125.2 | - | 7 | 2330 | ||
AK025600.1 | - | 8 | 1042 | |||
BC136629.1 | - | 9 | 1057 | 9020619 | ||
BC144323.1 | - | 9 | 1057 |