Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Penk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002927.2 | NP_001002927.1 | 2 | 1299 | ||
GenBank mRNA | AK135111.1 | - | 2 | 1041 | ||
AK161272.1 | - | 3 | 1155 | |||
BC049766.1 | - | 2 | 1299 | 6774387 | ||
M13227.1 | - | 2 | 913 | |||
M55181.1 | - | 2 | 1296 |