Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crtc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004062.2 | NP_001004062.1 | 14 | 3185 | ||
GenBank mRNA | AK129173.1 | - | 14 | 3182 | ||
BC056137.1 | - | 14 | 7773 | |||
BC068139.1 | - | 14 | 3191 | |||
BC080308.1 | - | 14 | 3185 | 5719562 |