Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145636.1 | NP_001139108.1 | 12 | 1420 | ||
XM_006710603.2 | XP_006710666.1 | 13 | 1718 | |||
XM_006710604.2 | XP_006710667.1 | 13 | 1706 | |||
XM_011541340.1 | XP_011539642.1 | 14 | 1842 | |||
XM_011541341.1 | XP_011539643.1 | 12 | 1454 | |||
XM_011541343.1 | XP_011539645.1 | 13 | 1578 | |||
XM_011541345.1 | XP_011539647.1 | 13 | 1652 | |||
XM_011541349.1 | XP_011539651.1 | 12 | 1388 | |||
XM_011541354.1 | XP_011539656.1 | 11 | 1356 | |||
XM_011541355.1 | XP_011539657.1 | 10 | 1123 | |||
XM_011541356.2 | XP_011539658.1 | 9 | 873 | |||
XM_017001140.1 | XP_016856629.1 | 13 | 1797 | |||
XM_017001141.1 | XP_016856630.1 | 12 | 1719 | |||
XM_017001142.1 | XP_016856631.1 | 11 | 1289 | |||
XM_017001143.1 | XP_016856632.1 | 10 | 1211 | |||
XM_017001148.1 | XP_016856637.1 | 10 | 1189 | |||
GenBank mRNA | AY254217.1 | - | 10 | 1040 | ||
BC026115.1 | - | 10 | 1854 | 4822241 |