Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300816.2 | NP_001287745.1 | 3 | 253 | ||
NM_001300817.2 | NP_001287746.1 | 3 | 253 | |||
NM_001300818.2 | NP_001287747.1 | 3 | 253 | |||
NM_001300819.2 | NP_001287748.1 | 2 | 187 | |||
NM_001300822.2 | NP_001287751.1 | 3 | 253 | |||
NM_001308127.1 | NP_001295056.1 | 3 | 283 | |||
NM_001308130.1 | NP_001295059.1 | 2 | 187 | |||
NM_017665.3 | NP_060135.1 | 2 | 187 | |||
NR_125337.2 | 2 | 191 | ||||
NR_131766.1 | 3 | 308 | ||||
NR_131767.1 | 2 | 191 | ||||
XM_011543502.1 | XP_011541804.1 | 3 | 282 | |||
XM_011543503.2 | XP_011541805.1 | 3 | 300 | |||
GenBank mRNA | AF086446.1 | - | 1 | 69 | ||
AK000101.1 | - | 2 | 131 | |||
AK122970.1 | - | 3 | 198 | |||
AK300543.1 | - | 2 | 155 | |||
BC005211.1 | - | 2 | 144 | 3961163 | ||
BC022443.1 | - | 3 | 194 | 4285562 | ||
BF983215.1 | - | 2 | 121 | |||
BG699335.1 | - | 3 | 233 | |||
BI826979.1 | - | 2 | 187 | |||
BM470321.1 | - | 2 | 138 | |||
BU165290.1 | - | 3 | 184 | |||
CB112418.1 | - | 3 | 194 | |||
DA466245.1 | - | 3 | 249 | |||
DA528172.1 | - | 2 | 131 | |||
DA905071.1 | - | 3 | 228 |