Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN2B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292038.1 | NP_001278967.1 | 13 | 2045 | ||
NM_015274.2 | NP_056089.1 | 13 | 2198 | |||
GenBank mRNA | AB023152.2 | - | 13 | 2157 | ||
AK025803.1 | - | 5 | 826 | |||
AK075246.1 | - | 1 | 136 | |||
AK301473.1 | - | 13 | 2030 | |||
AL832232.1 | - | 13 | 2168 | |||
AL833071.2 | - | 13 | 2171 | |||
BC033307.1 | - | 13 | 2167 | 4932398 | ||
BC036836.1 | - | 3 | 380 | |||
BC094773.1 | - | 13 | 2198 | 30531368 |