Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIMBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015347.4 | NP_056162.4 | 18 | 3232 | ||
XM_011538102.1 | XP_011536404.1 | 20 | 4050 | |||
XM_011538103.2 | XP_011536405.1 | 22 | 4512 | |||
XM_011538104.2 | XP_011536406.1 | 22 | 4327 | |||
XM_011538105.2 | XP_011536407.1 | 21 | 3784 | |||
XM_011538106.2 | XP_011536408.1 | 20 | 3625 | |||
XM_017019105.1 | XP_016874594.1 | 22 | 4525 | |||
XM_017019106.1 | XP_016874595.1 | 21 | 3784 | |||
XM_017019107.1 | XP_016874596.1 | 21 | 3606 | |||
XM_017019108.1 | XP_016874597.1 | 21 | 3789 | |||
XM_017019109.1 | XP_016874598.1 | 19 | 3333 | |||
XM_017019110.1 | XP_016874599.1 | 21 | 3606 | |||
XM_017019111.1 | XP_016874600.1 | 21 | 3606 | |||
GenBank mRNA | AB002316.1 | - | 18 | 3232 | ||
AY699979.1 | - | 15 | 2923 | |||
BX641152.1 | - | 5 | 7964 |