Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017554.2 | NP_060024.2 | 12 | 3930 | ||
XM_011512928.2 | XP_011511230.1 | 12 | 4021 | |||
GenBank mRNA | AB033094.1 | - | 7 | 1900 | ||
AK001770.1 | - | 8 | 958 | |||
AK026003.1 | - | 7 | 1759 | |||
AK304269.1 | - | 12 | 3898 | |||
AY134858.1 | - | 7 | 3103 | |||
BX648758.1 | - | 8 | 3249 | |||
DQ063584.1 | - | 9 | 3394 | |||
DQ063585.1 | - | 9 | 3394 |