Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256071.2 | NP_001243000.2 | 47, 48 | 12695 | ||
XM_005257545.4 | XP_005257602.2 | 48, 49 | 12883 | |||
XM_005257546.4 | XP_005257603.2 | 48, 49 | 12787 | |||
XM_006721995.3 | XP_006722058.1 | 48, 49 | 13170 | |||
XM_011525084.2 | XP_011523386.1 | 48, 49 | 12883 | |||
XM_017024905.1 | XP_016880394.1 | 43, 44 | 11750 | |||
GenBank mRNA | AB046774.1 | - | 7, 8 | 885 | ||
AB537889.1 | - | 47, 48 | 12688 | |||
AK025676.1 | - | 1, 2 | 101 | |||
AK074030.1 | - | 9, 10 | 1239 | |||
AL833201.1 | - | 7, 8 | 913 | |||
BX640932.1 | - | 13, 14 | 1851 | |||
BX647946.1 | - | 19, 20 | 3486 |