Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256071.2 | NP_001243000.2 | 55 | 13781 | ||
XM_005257545.4 | XP_005257602.2 | 56 | 13969 | |||
XM_005257546.4 | XP_005257603.2 | 56 | 13873 | |||
XM_006721995.3 | XP_006722058.1 | 56 | 14256 | |||
XM_011525084.2 | XP_011523386.1 | 56 | 13969 | |||
XM_017024905.1 | XP_016880394.1 | 51 | 12836 | |||
GenBank mRNA | AB046774.1 | - | 15 | 1971 | ||
AB537889.1 | - | 55 | 13774 | |||
AK025676.1 | - | 9 | 1187 | |||
AK025914.1 | - | 8 | 955 | |||
AK074030.1 | - | 17 | 2325 | |||
AK127358.1 | - | 21 | 2959 | |||
AL833201.1 | - | 15 | 1999 | |||
BX640932.1 | - | 21 | 2937 | |||
BX647946.1 | - | 27 | 4572 |