Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Helz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198298.1 | NP_938040.1 | 25 | 3708 | ||
GenBank mRNA | AB093209.1 | - | 26 | 3766 | ||
AK033094.1 | - | 9 | 1363 | |||
AK044226.1 | - | 10 | 1567 | |||
BC076626.1 | - | 25 | 3708 | 30640711 |