Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213564.3 | NP_998729.1 | 2 | 1886 | ||
XM_006256121.2 | XP_006256183.1 | 2 | 1874 | |||
XM_006256122.2 | XP_006256184.1 | 2 | 1819 | |||
GenBank mRNA | BC100649.1 | - | 1834 | 7932768 |